missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloronaphthalene, TRC

Product Code. 30137856
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137856 100 mg 100mg
17982705 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137856 Supplier Toronto Research Chemicals Supplier No. TRCD436670100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2050-69-3
Molecular Formula C10 H6 Cl2
Molecular Weight (g/mol) 197.06
Synonym Naphthalene, 1,2-dichloro-, 1,2-Dichloronaphthalene, NSC 90290, PCN 3, 1,2-DiCN, 1,2-DCN, PCN-3
IUPAC Name 1,2-dichloronaphthalene
SMILES Clc1ccc2ccccc2c1Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H6Cl2/c11-9-6-5-7-3-1-2-4-8(7)10(9)12/h1-6H
Formula Weight 195.9847
Chemical Name or Material 1,2-Dichloronaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.