missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-4-nitrobenzene, 99%, Thermo Scientific™

Product Code. 11433324
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11433324

Brand: Thermo Scientific Alfa Aesar A10620.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-54-7
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00007207
InChI Key NTBYINQTYWZXLH-UHFFFAOYSA-N
Synonym 3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech
PubChem CID 7443
IUPAC Name 1,2-dichloro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 39°C to 43°C
Boiling Point 255°C to 256°C
Flash Point 123°C (253°F)
Quantity 1000 g
UN Number UN3077
Beilstein 1818163
Formula Weight 192
Percent Purity 99%
Chemical Name or Material 1,2-Dichloro-4-nitrobenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.