Learn More
1,2-Dichloro-2-methylpropane, TRC
Product Code. 30011300
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011300 500 mg 500mg
30008219 100 mg 100mg
30005731 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011300 Supplier Toronto Research Chemicals Supplier No. TRCD437290500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 594-37-6
Molecular Formula C4H8Cl2
Molecular Weight (g/mol) 127.01
Synonym 1,2-Dichloroisobutane
SMILES CC(C)(Cl)CCl
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C4H8Cl2/c1-4(2,6)3-5/h3H2,1-2H3
Formula Weight 126.0003
Chemical Name or Material 1,2-Dichloro-2-methylpropane