Learn More
1,2-Dibromo-4,5-dimethylbenzene, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 24932-48-7 |
| Molecular Formula | C8H8Br2 |
| MDL Number | MFCD00082743 |
| Quantity | 100 g |
| SMILES | CC1=CC(Br)=C(Br)C=C1C |
| Molecular Weight (g/mol) | 263.96 |
| Percent Purity | 98% |
| Chemical Name or Material | 1,2-Dibromo-4,5-dimethylbenzene |