missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diaminocyclohexanetetraacetic acid monohydrate, Solstice
Description
Solstice's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Quantity | 6 x 25 g |
| UN Number | NONH for all modes of transport |
| Formula Weight | 364.35 |
| Percent Purity | ≥98.5% |
| Chemical Name or Material | 1,2-Diaminocyclohexanetetraacetic acid monohydrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?