missing translation for 'onlineSavingsMsg'
Learn More

1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride, 98%, Thermo Scientific™

Product Code. p-7056107
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439206 5 g 5g
15449206 25 g 25g
15459206 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15439206 Supplier Thermo Scientific Alfa Aesar Supplier No. H64862.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130198-05-9
Molecular Formula C15H24ClNO2
Molecular Weight (g/mol) 285.812
MDL Number MFCD06658143
InChI Key NTKXIDDUCSFBBF-UHFFFAOYSA-N
Synonym 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride, 1-2-amino-1-4-methoxyphenyl-ethyl-cyclohexanol hydrochloride, 2-4-methoxyphenyl-2-1-hydroxycyclohexyl ethylamine hydrochloride, 1-4-methoxyphenyl-2-aminoethyl cyclohexanol hydrochloride, n,n-didesmethylvenlafaxine hydrochloride, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexan-1-ol hydrochloride, 1-2-amino-1-4-methyoxyphenyl ethyl cyclohexanol.hcl, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol.hcl, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hcl
PubChem CID 9795856
IUPAC Name 1-[2-amino-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride
SMILES COC1=CC=C(C=C1)C(CN)C2(CCCCC2)O.Cl

Specifications

Melting Point 174°C to 178°C (literature)
Color White
Quantity 5 g
Beilstein 4215444
Formula Weight 285.81
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.