Learn More
1-(2,6-Dichlorophenyl)indolin-2,3-dione, Mikromol™
Product Code. 30052375
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052375 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052375 Supplier Mikromol Supplier No. MM0006.17
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(2,6-Dichlorophenyl)indolin-2,3-dione
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(2,6-Dichlorophenyl)indolin-2,3-dione
CAS 24542-74-3
Analyte or Component Names 1-(2,6-Dichlorophenyl)indolin-2,3-dione
Shipping Condition Room Temperature
Molecular Formula C14 H7 Cl2 N O2
Certifications/Compliance ISO 9001
Synonym 1-(2,6-Dichlorophenyl)indolin-2,3-dione
InChI Formula InChI=1S/C14H7Cl2NO2/c15-9-5-3-6-10(16)12(9)17-11-7-2-1-4-8(11)13(18)14(17)19/h1-7H
SMILES Clc1cccc(Cl)c1N2C(=O)C(=O)c3ccccc23
IUPAC Name 1-(2,6-dichlorophenyl)indole-2,3-dione
Molecular Weight (g/mol) 292.12
Quantity 100 mg
Formula Weight 290.9854 g/mol