Learn More
1-(2,4,6-Trimethoxybenzyl)piperazine Dihydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2,4,6-Trimethoxybenzyl)piperazine Dihydrochloride |
| CAS | 113698-83-2 |
| Molecular Formula | C14 H22 N2 O3 . 2 Cl H |
| Quantity | 100 mg |
| Synonym | Piperazine','1-[(2,4,6-trimethoxyphenyl)methyl]-','hydrochloride (1:2), Piperazine','1-[(2,4,6-trimethoxyphenyl)methyl]-','dihydrochloride (9CI), 1-(2,4,6-Trimethoxybenzyl)piperazine Dihydrochloride, Trimetazidine Dihydrochloride Imp. F (EP) as Dihydrochloride |
| InChI Formula | InChI=1S/C14H22N2O3.2ClH/c1-17-11-8-13(18-2)12(14(9-11)19-3)10-16-6-4-15-5-7-16;;/h8-9,15H,4-7,10H2,1-3H3;2*1H |
| SMILES | Cl.Cl.COc1cc(OC)c(CN2CCNCC2)c(OC)c1 |
| IUPAC Name | 1-[(2,4,6-trimethoxyphenyl)methyl]piperazine;dihydrochloride |
| Molecular Weight (g/mol) | 339.26 |
| Formula Weight | 338.1164 |