missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,2,3-Trichloro-4-nitrobenzene, 97%

Product Code. 11499176
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499176 10 g 10g
11409186 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499176 Supplier Alfa Aesar™ Supplier No. A18038.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 17700-09-3
Molecular Formula C6H2Cl3NO2
Molecular Weight (g/mol) 226.437
MDL Number MFCD00007063
InChI Key BGKIECJVXXHLDP-UHFFFAOYSA-N
Synonym 2,3,4-trichloronitrobenzene, trichloronitrobenzene, 2,3,4-trichloro-1-nitrobenzene, 4-nitro-1,2,3-trichlorobenzene, benzene, 1,2,3-trichloro-4-nitro, benzene, 4-nitro-1,2,3-trichloro, 1,2,3-trichloronitrobenzene, 1,2,3-tris chloranyl-4-nitro-benzene, pubchem8437, acmc-1bpdn
PubChem CID 28697
IUPAC Name 1,2,3-trichloro-4-nitrobenzene
SMILES C1=CC(=C(C(=C1[N+](=O)[O-])Cl)Cl)Cl

Specifications

Melting Point 53°C to 56°C
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN2811
Beilstein 2211951
Formula Weight 226.45
Percent Purity 97%
Chemical Name or Material 1,2,3-Trichloro-4-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.