Learn More
1-(2,3-Dichlorophenyl)ethanone, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2,3-Dichlorophenyl)ethanone |
| CAS | 56041-57-7 |
| Molecular Formula | C8H6OCl2 |
| Quantity | 50 mg |
| Synonym | 2',3'-Dichloroacetophenone |
| InChI Formula | InChI=1S/C8H6Cl2O/c1-5(11)6-3-2-4-7(9)8(6)10/h2-4H,1H3 |
| SMILES | CC(=O)c1cccc(Cl)c1Cl |
| IUPAC Name | 1-(2,3-dichlorophenyl)ethanone |
| Molecular Weight (g/mol) | 189.04 |
| Formula Weight | 187.9796 |