Learn More
1,2,3,6-Tetrahydro-Alpha,4-bis(p-methoxyphenyl)-1-methyl-3-pyridinemethanol, TRC
Product Code. 30411103
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411103 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30411103 Supplier Toronto Research Chemicals Supplier No. TRCT11759050MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2,3,6-Tetrahydro-alpha,4-bis(p-methoxyphenyl)-1-methyl-3-pyridinemethanol
CAS 10185-04-3
Molecular Formula C21H25NO3
Quantity 50 mg
Synonym 1,2,3,6-Tetrahydro-α,4-bis(4-methoxyphenyl)-1-methyl-3-pyridinemethanol
InChI Formula InChI=1S/C21H25NO3/c1-22-13-12-19(15-4-8-17(24-2)9-5-15)20(14-22)21(23)16-6-10-18(25-3)11-7-16/h4-12,20-21,23H,13-14H2,1-3H3
SMILES OC(C1C(C2=CC=C(OC)C=C2)=CCN(C1)C)C3=CC=C(C=C3)OC
IUPAC Name (4-methoxyphenyl)(4-(4-methoxyphenyl)-1-methyl-1,2,3,6-tetrahydropyridin-3-yl)methanol
Molecular Weight (g/mol) 399.44
Formula Weight 399.18