missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,6-Tetragalloylglucose, MedChemExpress

Product Code. 16417257
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16417257 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16417257 Supplier Medchem Express Supplier No. HY111832

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 1,2,3,6-Tetragalloylglucose is a potent UDP glucuronosyltransferase 1 family, polypeptide A1 (UGT1A1) inhibitor, with a Ki of 1.68 μM.

1,2,3,6-Tetragalloylglucose is a potent UDP glucuronosyltransferase 1 family, polypeptide A1 (UGT1A1) inhibitor, with a Ki of 1.68 μM.

Specifications

Chemical Name or Material 1,2,3,6-Tetragalloylglucose
CAS 79886-50-3
Color White
Purity Grade Notes Research
Molecular Formula C34H28O22
Quantity 5 mg
Synonym TeGG
Solubility Information DMSO : 100 mg/mL (126.81 mM; Need ultrasonic)
SMILES O=C(C1=CC(O)=C(O)C(O)=C1)O[C@@H]2[C@H]([C@@H]([C@@H](COC(C3=CC(O)=C(O)C(O)=C3)=O)O[C@H]2OC(C4=CC(O)=C(O)C(O)=C4)=O)O)OC(C5=CC(O)=C(O)C(O)=C5)=O
Molecular Weight (g/mol) 788.57
Formula Weight 788.57
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.