Learn More
1,2,3,4-Tetrahydro-1,2-dimethyl-4,6-isoquinolinediol, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2,3,4-Tetrahydro-1,2-dimethyl-4,6-isoquinolinediol |
| CAS | 102830-16-0 |
| Molecular Formula | C11 H15 N O2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C11H15NO2/c1-7-9-4-3-8(13)5-10(9)11(14)6-12(7)2/h3-5,7,11,13-14H,6H2,1-2H3 |
| SMILES | CC1N(C)CC(O)c2cc(O)ccc12 |
| IUPAC Name | 1,2-dimethyl-3,4-dihydro-1H-isoquinoline-4,6-diol |
| Molecular Weight (g/mol) | 193.242 |
| Formula Weight | 193.11 |