missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Cyclobutanetetracarboxylic Acid 1,2,3,4-Tetramethyl Ester, TRC

Product Code. 30134573
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134573 250 mg 250mg
30010244 100 mg 100mg
30010039 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134573 Supplier Toronto Research Chemicals Supplier No. TRCC989408250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14495-41-1
Molecular Formula C12 H16 O8
Molecular Weight (g/mol) 288.25
Synonym 1,2,3,4-Cyclobutanetetracarboxylic acid, 1,2,3,4-tetramethyl ester, 1,2,3,4-Cyclobutanetetracarboxylic acid, tetramethyl ester (6CI,7CI,8CI,9CI), Tetramethyl cyclobutane-1,2,3,4-tetracarboxylate
IUPAC Name tetramethyl cyclobutane-1,2,3,4-tetracarboxylate
SMILES COC(=O)C1C(C(C1C(=O)OC)C(=O)OC)C(=O)OC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H16O8/c1-17-9(13)5-6(10(14)18-2)8(12(16)20-4)7(5)11(15)19-3/h5-8H,1-4H3
Formula Weight 288.0845
Percent Purity >95%
Chemical Name or Material 1,2,3,4-Cyclobutanetetracarboxylic Acid 1,2,3,4-Tetramethyl Ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.