Learn More
1-(1-Naphthyloxy)-2,3-epoxypropane, Mikromol™
Product Code. 30051632
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051632 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30051632 Supplier Mikromol Supplier No. MM0043.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(1-Naphthyloxy)-2,3-epoxypropane
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(1-Naphthyloxy)-2,3-epoxypropane
CAS 2461-42-9
Analyte or Component Names 1-(1-Naphthyloxy)-2,3-epoxypropane
Shipping Condition Room Temperature
Molecular Formula C13 H12 O2
Certifications/Compliance ISO 17025
Synonym 1-(1-Naphthyloxy)-2,3-epoxypropane
InChI Formula InChI=1S/C13H12O2/c1-2-6-12-10(4-1)5-3-7-13(12)15-9-11-8-14-11/h1-7,11H,8-9H2
SMILES C(Oc1cccc2ccccc12)C3CO3
IUPAC Name 2-(naphthalen-1-yloxymethyl)oxirane
Molecular Weight (g/mol) 200.23
Quantity 100 mg
Formula Weight 200.0837 g/mol