Learn More
1,1-Iminodi-2-propanol, Sigma-Aldrich™
Product Code. 32238870
Change view
Click to view available options
Quantity:
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32238870 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238870 Supplier Sigma Aldrich Supplier No. 8032649025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1-Iminodi-2-propanol
CAS 110-97-4
Density 0.99 g/cm³ at 20°C
Boiling Point 249°C (1013 hPa)
Flash Point 135°C (closed cup)
Melting Range 40.0°C to 48.0°C
Molecular Formula [CH3CH(OH)CH2]2NH
Quantity 25 kg
Synonym 1,1-Iminodi-2-propanol, Diisopropanolamine, Bis(2-hydroxypropyl)amine, 1,1-Iminodi-2-propanol
Molecular Weight (g/mol) 133.19
Grade Not Available