Learn More
1,1-Iminodi-2-propanol, Sigma-Aldrich™
Product Code. 15894121
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894121 100 g 100g
15804131 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15894121 Supplier Sigma Aldrich Supplier No. 8032640100
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-97-4
Grade Research
Molecular Formula [CH3CH(OH)CH2]2NH
Molecular Weight (g/mol) 133.19
Specifications
Melting Point 40.0°C to 48.0°C
Density 0.99 g/cm3 at 20°C
Boiling Point 249°C/1013 hPa
MDL Number MFCD00004531
Quantity 100 g
Synonym 1,1-Iminodi-2-propanol, Diisopropanolamine, Bis(2-hydroxypropyl)amine, 1,1-Iminodi-2-propanol