Learn More
1-(1-Ethylpiperidin-3-yl)methanamine, TRC
Product Code. 30330658
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330658 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30330658 Supplier Toronto Research Chemicals Supplier No. TRCB434335100MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(1-Ethylpiperidin-3-yl)methanamine
CAS 102459-02-9
Molecular Formula C8H18N2
Quantity 100 mg
Synonym [(1-Ethylpiperidin-3-yl)methyl]amine
InChI Formula InChI=1S/C8H18N2/c1-2-10-5-3-4-8(6-9)7-10/h8H,2-7,9H2,1H3
SMILES CCN1CCCC(CN)C1
IUPAC Name (1-ethylpiperidin-3-yl)methanamine
Molecular Weight (g/mol) 142.24
Formula Weight 142.147