Learn More
[1,1'-Biphenyl]-2-carboxylic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 947-84-2 |
| Molecular Formula | C13H10O2 |
| MDL Number | MFCD00002463 |
| Quantity | 1 kg |
| SMILES | C1=CC=CC(=C1C2=CC=CC=C2)C(O)=O |
| Molecular Weight (g/mol) | 198.22 |
| Percent Purity | 98% |
| Chemical Name or Material | [1,1'-Biphenyl]-2-carboxylic acid |