Learn More
1,1,3,3,5,5,7,7,9,9-Decamethylpentasiloxane, TRC
Specifications
Specifications
| Chemical Name or Material | 1,1,3,3,5,5,7,7,9,9-Decamethylpentasiloxane |
| CAS | 995-83-5 |
| Molecular Formula | C10 H32 O4 Si5 |
| Quantity | 2.5 g |
| Synonym | 1,9-Dihydridodecamethylpentasiloxane, 1,9-Dihydrodecamethylpentasiloxane, 1,9-Dihydrogendecamethylpentasiloxane |
| InChI Formula | InChI=1S/C10H32O4Si5/c1-15(2)11-17(5,6)13-19(9,10)14-18(7,8)12-16(3)4/h15-16H,1-10H3 |
| SMILES | C[SiH](C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[SiH](C)C |
| IUPAC Name | bis[[dimethylsilyloxy(dimethyl)silyl]oxy]-dimethylsilane |
| Molecular Weight (g/mol) | 356.786 |
| Formula Weight | 356.115 |