Learn More
1,1,3,3,3-Pentafluoro-2-(fluoromethoxy)-1-propene, TRC
Product Code. 30009170
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009170 5 mg 5mg
30008262 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009170 Supplier Toronto Research Chemicals Supplier No. TRCP2272455MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 58109-34-5
Molecular Formula C4 H2 F6 O
Molecular Weight (g/mol) 180.05
Synonym 1-Propene, 1,1,3,3,3-pentafluoro-2-(fluoromethoxy)-, 1,1,3,3,3-Pentafluoro-2-(fluoromethoxy)-1-propene, 2-(Fluoromethoxy)-1,1,3,3,3-pentafluoro-1-propene, Compound A, Fluoromethyl 2,2-difluoro-1-(trifluoromethyl)vinyl ether
IUPAC Name 1,1,3,3,3-pentafluoro-2-(fluoromethoxy)prop-1-ene
SMILES FCOC(=C(F)F)C(F)(F)F
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C4H2F6O/c5-1-11-2(3(6)7)4(8,9)10/h1H2
Formula Weight 180.001
Chemical Name or Material 1,1,1,3,3-Pentafluoroisopropenyl fluoromethyl ether