missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,1,1-Tris(chloromethyl)ethane, 98%

Product Code. 11468127
Change view
Click to view available options
Quantity:
25g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468127 25g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11468127 Supplier Alfa Aesar™ Supplier No. A17514.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1067-09-0
Boiling Point 85°C to 87°C (14mmHg)
Flash Point 86°C (187°F)
Molecular Formula C5H9Cl3
MDL Number MFCD00013686
Quantity 25g
Synonym 1,1,1-tris chloromethyl ethane, 1,3-dichloro-2-chloromethyl-2-methylpropane, 2-chloromethyl-1,3-dichloro-2-methylpropane, acmc-20ap8n, pentane, 1,1,1-trichloro, 1-chloro-2,2-bis chloromethyl propane, 1,3-dichloro-2-chloromethyl-2-methylpropane #, propane,1,3-dichloro-2-chloromethyl-2-methyl
InChI Key BYXOMFFBGDPXHB-UHFFFAOYSA-N
SMILES CC(CCl)(CCl)CCl
IUPAC Name 1,3-dichloro-2-(chloromethyl)-2-methylpropane
Molecular Weight (g/mol) 175.477
PubChem CID 136826
Formula Weight 175.49
Percent Purity 98%
Chemical Name or Material 1,1,1-Tris(chloromethyl)ethane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.