Learn More
1,1,1-Trimethoxybutane, TRC
Specifications
Specifications
| Chemical Name or Material | 1,1,1-Trimethoxybutane |
| CAS | 43083-12-1 |
| Molecular Formula | C7H16O3 |
| Quantity | 25 g |
| Synonym | Trimethyl Ester Orthobutyric Acid, 1,1,1-Trimethoxybutane, Methyl Orthobutyrate, Trimethyl Orthobutanoate, Trimethyl Orthobutyrate |
| InChI Formula | InChI=1S/C7H16O3/c1-5-6-7(8-2,9-3)10-4/h5-6H2,1-4H3 |
| SMILES | CCCC(OC)(OC)OC |
| IUPAC Name | 1,1,1-trimethoxybutane |
| Molecular Weight (g/mol) | 148.2 |
| Formula Weight | 148.1099 |