Learn More
1,1,1-Trifluoromethanesulfonic Acid 2-(2-Propen-1-yl)phenyl Ester-d4, TRC
Product Code. 30455714
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455714 100 mg 100mg
30452077 5 mg 5mg
30454043 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30455714 Supplier Toronto Research Chemicals Supplier No. TRCT790547100MG
 Restricted Item
Estimated Shipment: 12-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1,1-Trifluoromethanesulfonic Acid 2-(2-Propen-1-yl)phenyl Ester-d4
Molecular Formula C10 D4 H5 F3 O3 S
Quantity 100 mg
Synonym Trifluoromethanesulfonic Acid 2-(2-Propen-1-yl)phenyl Ester-d4
InChI Formula InChI=1S/C10H9F3O3S/c1-2-5-8-6-3-4-7-9(8)16-17(14,15)10(11,12)13/h2-4,6-7H,1,5H2/i3D,4D,6D,7D
SMILES [2H]c1c([2H])c([2H])c(OS(=O)(=O)C(F)(F)F)c(CC=C)c1[2H]
IUPAC Name (2,3,4,5-tetradeuterio-6-prop-2-enylphenyl) trifluoromethanesulfonate
Molecular Weight (g/mol) 270.262
Formula Weight 270.048