Learn More
1,1,1,3,3,3-Hexafluoro-2-propanol, MilliporeSigma™
Product Code. 15876380
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15876380 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 15876380 Supplier Sigma Aldrich Supplier No. 8045150100
 Restricted Item
Estimated Shipment: 19-11-2026
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 920-66-1
Melting Point 3°C
Color Colorless
Density 1.62g/cm3 (20°C)
pH 3 to 4 (aq. soln.)
Boiling Point 58°C
Flash Point >100°C (>212°F)
Odor Characteristic
Packaging Glass Bottle
Molecular Formula C3H2F6O
Refractive Index 1.2752 (25°C, 589nm)
Quantity 100 mL
UN Number UN 3265
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
Solubility Information Water: soluble at 20°C (68°F)
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
SMILES C(C(F)(F)F)(C(F)(F)F)O
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular Weight (g/mol) 168.038
PubChem CID 13529
ChEBI CHEBI:63104
Formula Weight 168.03
Vapor Pressure 269 hPa (30°C)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-hexafluoro-2-propanol