missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,2-Tetrachloroethane, TRC

Product Code. 30180059
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180059 1 g 1g
30179789 5 g 5g
30180039 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30180059 Supplier Toronto Research Chemicals Supplier No. TRCT2838801G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-20-6
Molecular Formula C2 H2 Cl4
Molecular Weight (g/mol) 167.85
Synonym 1,1,1,2-Tetrachloroethane, (Chloromethyl)trichloromethane, F 130a, HCC 130a, TCA, Tetrachloroethane
IUPAC Name 1,1,1,2-tetrachloroethane
SMILES ClCC(Cl)(Cl)Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H2Cl4/c3-1-2(4,5)6/h1H2
Formula Weight 165.8911
Chemical Name or Material 1,1,1,2-Tetrachloroethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.