missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2a Kappa Isotype Control (M2AK) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30279268 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30279268 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30279268 Supplier Novus Biologicals Supplier No. NBP196981MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgG2a Kappa Monoclonal antibody specifically detects IgG2a Kappa in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, SDS-Page, Fluorophore-Linked Immunosorbent Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2a Kappa
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page, Fluorophore-Linked Immunosorbent Assay
Classification Monoclonal
Clone M2AK
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, SDS-Page, Fluorophore-Linked Immunosorbent Assay
Formulation 50 mM Sodium Borate
Host Species Mouse
Purification Method Protein A purified
Quantity 0.5 mL
Regulatory Status RUO
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.