Learn More
m-Toluidine-d7 (NH2), CDN
Specifications
Specifications
| Chemical Name or Material | 3-Methylaniline-D7 |
| CAS | 68408-25-3 |
| Molecular Formula | C7 D7 H2 N |
| Quantity | 0.5 g |
| Synonym | Benzen-2,3,4,6-d4-amine, 5-(methyl-d3)- (9 CI, ACI), 5-(Methyl-d3)benzen-2,3,4,6-d4-amine (ACI), 2,3,4,6-Tetradeuterio-5-trideuteriomethyl-aniline, 3-Methylaniline-d7, 3-Methylaniline-D7, m-Toluidine-d7 (NH2) |
| InChI Formula | InChI=1 S/C7H9N/c1-6-3-2-4-7(8)5-6/h2-5 H,8H2,1H3/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(N)c([2 H])c(c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(trideuteriomethyl)aniline |
| Molecular Weight (g/mol) | 114.196 |
| Formula Weight | 114.117 g/mol |
| Show More |