Learn More
1-Bromo-2,6-difluorobenzene-d3, CDN
Product Code. 30321336
Change view
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321336 0.5 g 0.5g
30317128 0.25 g 0.25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321336 Supplier CDN Supplier No. CDND60640.5G
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Bromo-2,6-difluorobenzene-3,4,5-D6
CAS 1219803-73-2
Molecular Formula C6 D3 Br F2
Quantity 0.5 g
Synonym 2-Bromo-1,3-difluorobenzene-4,5,6-d3, 1-Bromo-2,6-difluorobenzene-3,4,5-D6, 2,6-Difluoro-1-bromobenzene-d3, 2,6-Difluorobromobenzene-d3
InChI Formula InChI=1 S/C6H3BrF2/c7-6-4(8)2-1-3-5(6)9/h1-3 H/i1D,2 D,3 D
SMILES [2 H]c1c([2 H])c(F)c(Br)c(F)c1[2 H]
IUPAC Name 1-bromo-3,4,5-trideuterio-2,6-difluorobenzene
Molecular Weight (g/mol) 196.0073
Formula Weight 194.9574 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity