missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GPR12 Monoclonal antibody specifically detects GPR12 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry
Specifications
Specifications
| Antigen | GPR12 |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 584940 |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ18149, FLJ97704, G protein-coupled receptor 12, GPCR12, GPCR21, G-protein coupled receptor 12, MGC138349, MGC138351 |
| Host Species | Mouse |
| Immunogen | NS0 mouse myeloma cell line transfected with human GPR12, Met1-Val334, Accession # P47775 |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?