missing translation for 'onlineSavingsMsg'
Learn More

Craniofacial Development Protein 1 Antibody [Janelia Fluor™ 635], Novus Biologicals™

Product Code. 30170038
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30170038 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30170038 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP318436JF635

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Craniofacial Development Protein 1 Polyclonal antibody specifically detects Craniofacial Development Protein 1 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Craniofacial Development Protein 1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 635
Formulation 50mM Sodium Borate
Gene Alias BCNTBUCENTAUR, Bucentaur, CP27phosphoprotein (Bucentaur), craniofacial development protein 1, p97, SWC5, Yeti
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 249 and 299..
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10428
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.