missing translation for 'onlineSavingsMsg'
Learn More

Carboxypeptidase A1/CPA1 Antibody (rCPA1/8688), PE, Novus Biologicals™

Product Code. 18602785 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18602785 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18602785 Supplier Novus Biologicals Supplier No. NBP321082PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Carboxypeptidase A1/CPA1 Monoclonal antibody specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase A1/CPA1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rCPA1/8688
Conjugate PE
Formulation PBS
Gene Alias carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1
Host Species Mouse
Immunogen Recombinant full-length human Carboxypeptidase A1/CPA1 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mast Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1357
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.