Learn More
Carboxytolbutamide-d9 (butyl-d9), CDN
Specifications
Specifications
| Chemical Name or Material | 4-Carboxy Tolbutamide-d9 |
| CAS | 1219802-95-5 |
| Molecular Formula | C12 D9 H7 N2 O5 S |
| Quantity | 0.005 g |
| Synonym | 4-(1,1,2,2,3,3,4,4,4-Nonadeuteriobutylcarbamoylsulfamoyl)benzoic acid, 4-[[[(Butyl-d9-amino)carbonyl]amino]sulfonyl]benzoic acid, 1-(Butyl-d9)-3-(p-carboxyphenyl)sulfonylurea, Carboxycarbutamide-d9, Carboxytolbutamide-d9, Tolbutamide carboxylic acid-d9, 4-Carboxytolbutamide-d9, 4-Carboxytolbutamide-D9 (butyl-D9) |
| InChI Formula | InChI=1 S/C12H16N2O5S/c1-2-3-8-13-12(17)14-20(18,19)10-6-4-9(5-7-10)11(15)16/h4-7 H,2-3,8H2,1H3,(H,15,16)(H2,13,14,17)/i1D3,2D2,3D2,8D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])NC(=O)NS(=O)(=O)c1ccc(cc1)C(=O)O |
| IUPAC Name | 4-(1,1,2,2,3,3,4,4,4-nonadeuteriobutylcarbamoylsulfamoyl)benzoic acid |
| Molecular Weight (g/mol) | 309.3863 |
| Formula Weight | 309.1345 g/mol |
| Show More |