Thermo Scientific™ Pierce™ Mercaptoethylamine-HCl
Pure, crystalline 2-aminoethanethiol (2-MEA-HCl, also called cysteamine-HCl), CAS 156-57-0; selectively reduces antibody hinge-region disulfide bonds.
Additional Details:
Additional Details: CAS Number: 156-57-0 Weight: 0.80000kg
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 11816784
Quantity Price
1 325.0  EUR / Pack of 6
In stock, ready to ship: 11
Add to basket
Description and Specification
Specification
Content And Storage Store desiccated at 4°C. Product is shipped at ambient temperature.
Product Line Pierce
Form Solid
Quantity 6 x 6 mg
Product Type Mercaptoethylamine Hydrochloride
M.W. 113.61
Safety and Handling
[2-MERCAPTOETHYLAMINE - HCL]
[Warning]
[{code=H302, sequence=1, description=Harmful if swallowed.}, {code=H315, sequence=2, description=Causes skin irritation.}, {code=H319, sequence=3, description=Causes serious eye irritation.}, {code=H335, sequence=4, description=May cause respiratory irritation.}]
[{code=P261, sequence=51, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P264, sequence=52, description=Wash hands thoroughly after handling.}, {code=P270, sequence=53, description=Do not eat, drink or smoke when using this product.}, {code=P271, sequence=54, description=Use only outdoors or in a well-ventilated area.}, {code=P280, sequence=55, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P312, sequence=56, description=IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.}, {code=P302+P352, sequence=57, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P304+P340, sequence=58, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=59, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P312, sequence=60, description=Call a POISON CENTER/doctor/if you feel unwell.}, {code=P330, sequence=61, description=Rinse mouth.}, {code=P332+P313, sequence=62, description=If skin irritation occurs: Get medical advice/attention.}, {code=P337+P313, sequence=63, description=If eye irritation persists: Get medical advice/attention.}, {code=P362+P364, sequence=64, description=Take off contaminated clothing and wash it before reuse.}, {code=P501b, sequence=65, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: 2-MercaptoethylamineHydrochloride}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Product Identifier
  • 2-MERCAPTOETHYLAMINE - HCL
Signal Word
  • Warning
Hazard Statement
  • H302- Harmful if swallowed.
  • H315- Causes skin irritation.
  • H319- Causes serious eye irritation.
  • H335- May cause respiratory irritation.
Precautionary Statement
  • P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264- Wash hands thoroughly after handling.
  • P270- Do not eat, drink or smoke when using this product.
  • P271- Use only outdoors or in a well-ventilated area.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312- IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352- IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312- Call a POISON CENTER/doctor/if you feel unwell.
  • P330- Rinse mouth.
  • P332+P313- If skin irritation occurs: Get medical advice/attention.
  • P337+P313- If eye irritation persists: Get medical advice/attention.
  • P362+P364- Take off contaminated clothing and wash it before reuse.
  • P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
  • MIXTURE LIST- Contain: 2-MercaptoethylamineHydrochloride