Nalpha-Boc-L-lysine, 97%
CAS: 13734-28-6 | C11H22N2O4 | 246.31 g/mol
Additional Details:
Additional Details: CAS Number: 13734-28-6 Weight: 0.10000kg
Quantity 100 g
Disclaimers: RUO – Research Use Only
Product Code. 15400799
Quantity Price
1 954.0  EUR / 100g
Estimated Shipment
21-08-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
??product.attributeSet.Chemical data??
CAS 13734-28-6
Molecular Formula C11H22N2O4
Molecular Weight (g/mol) 246.31
MDL Number MFCD00038203
InChI Key DQUHYEDEGRNAFO-SVGMAFHSNA-N
Synonym boc-lys-oh,boc-lysine,n-boc-l-lysine,boc-l-lysine,s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid,nalpha-tert-butoxycarbonyl-l-lysine,n-alpha-tert-butoxycarbonyl-l-lysine,nalpha-boc-l-lysine,boc-lys,n alpha-boc-l-lysine
PubChem CID 2733284
IUPAC Name (2S)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O
Specification
Chemical Name or Material Nalpha-Boc-L-lysine
Melting Point ∼205°C (decomposition)
CAS 13734-28-6
Color White
Molecular Formula C11H22N2O4
MDL Number MFCD00038203
Beilstein 4252546
Synonym boc-lys-oh,boc-lysine,n-boc-l-lysine,boc-l-lysine,s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid,nalpha-tert-butoxycarbonyl-l-lysine,n-alpha-tert-butoxycarbonyl-l-lysine,nalpha-boc-l-lysine,boc-lys,n alpha-boc-l-lysine
Synonym boc-lys-oh,boc-lysine,n-boc-l-lysine,boc-l-lysine,s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid,nalpha-tert-butoxycarbonyl-l-lysine,n-alpha-tert-butoxycarbonyl-l-lysine,nalpha-boc-l-lysine,boc-lys,n alpha-boc-l-lysine
Synonym boc-lys-oh,boc-lysine,n-boc-l-lysine,boc-l-lysine,s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid,nalpha-tert-butoxycarbonyl-l-lysine,n-alpha-tert-butoxycarbonyl-l-lysine,nalpha-boc-l-lysine,boc-lys,n alpha-boc-l-lysine
Synonym boc-lys-oh,boc-lysine,n-boc-l-lysine,boc-l-lysine,s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid,nalpha-tert-butoxycarbonyl-l-lysine,n-alpha-tert-butoxycarbonyl-l-lysine,nalpha-boc-l-lysine,boc-lys,n alpha-boc-l-lysine
Synonym boc-lys-oh,boc-lysine,n-boc-l-lysine,boc-l-lysine,s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid,nalpha-tert-butoxycarbonyl-l-lysine,n-alpha-tert-butoxycarbonyl-l-lysine,nalpha-boc-l-lysine,boc-lys,n alpha-boc-l-lysine
Synonym boc-lys-oh,boc-lysine,n-boc-l-lysine,boc-l-lysine,s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid,nalpha-tert-butoxycarbonyl-l-lysine,n-alpha-tert-butoxycarbonyl-l-lysine,nalpha-boc-l-lysine,boc-lys,n alpha-boc-l-lysine
Synonym boc-lys-oh,boc-lysine,n-boc-l-lysine,boc-l-lysine,s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid,nalpha-tert-butoxycarbonyl-l-lysine,n-alpha-tert-butoxycarbonyl-l-lysine,nalpha-boc-l-lysine,boc-lys,n alpha-boc-l-lysine
Synonym boc-lys-oh,boc-lysine,n-boc-l-lysine,boc-l-lysine,s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid,nalpha-tert-butoxycarbonyl-l-lysine,n-alpha-tert-butoxycarbonyl-l-lysine,nalpha-boc-l-lysine,boc-lys,n alpha-boc-l-lysine
Synonym boc-lys-oh,boc-lysine,n-boc-l-lysine,boc-l-lysine,s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid,nalpha-tert-butoxycarbonyl-l-lysine,n-alpha-tert-butoxycarbonyl-l-lysine,nalpha-boc-l-lysine,boc-lys,n alpha-boc-l-lysine
Synonym boc-lys-oh,boc-lysine,n-boc-l-lysine,boc-l-lysine,s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid,nalpha-tert-butoxycarbonyl-l-lysine,n-alpha-tert-butoxycarbonyl-l-lysine,nalpha-boc-l-lysine,boc-lys,n alpha-boc-l-lysine
Solubility Information Soluble in water. Slightly soluble in methanol.
InChI Key DQUHYEDEGRNAFO-SVGMAFHSNA-N
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O
IUPAC Name (2S)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
Molecular Weight (g/mol) 246.31
Quantity 100 g
PubChem CID 2733284
Formula Weight 246.3
Percent Purity 97%
Physical Form Powder
Applications
N-Boc-L-lysine is a protected analogue of a major amino acid participating in the binding of AcH to proteins. N-Boc-L-lysine as well as other t-butoxycarbonyl (Boc) amino acids can be converted to their respective amino acids via cleavage of the urethane bond.
Solubility
Soluble in water. Slightly soluble in methanol.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.