D(-)-Pantolactone, 99%
CAS: 599-04-2 | C6H10O3 | 130.14 g/mol
Product Details
79.88 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Additional Details:
Additional Details: CAS Number: 599-04-2 Weight: 0.02500kg
Quantity 25 g
Packaging Glass bottle
Disclaimers: RUO – Research Use Only
Product Code. 10696032
Quantity Price
1 80.7  EUR / 25g
In stock, ready to ship: 3
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
??product.attributeSet.Chemical data??
CAS 599-04-2
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00005392
InChI Key SERHXTVXHNVDKA-BYPYZUCNSA-N
Synonym d---pantolactone,r-pantolactone,d-pantolactone,r-pantoyl lactone,d---pantoyl lactone,pantothenic lactone,r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one,unii-j288d7o0js,2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r,r---pantolactone
PubChem CID 439368
ChEBI CHEBI:16719
IUPAC Name (3R)-3-hydroxy-4,4-dimethyloxolan-2-one
SMILES CC1(COC(=O)C1O)C
Specification
Chemical Name or Material D(-)-Pantolactone
CAS 599-04-2
Melting Point 88.0°C to 92.0°C
Color White
Boiling Point 120.0°C to 122.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H10O3
MDL Number MFCD00005392
Quantity 25 g
Beilstein 18, V,1, 22
Fieser 16,269; 17,270
Merck Index 15, 7116
Specific Rotation Condition −50.50° (25°C c=2,H2O)
Specific Rotation − 50.50
Synonym d---pantolactone,r-pantolactone,d-pantolactone,r-pantoyl lactone,d---pantoyl lactone,pantothenic lactone,r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one,unii-j288d7o0js,2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r,r---pantolactone
Synonym d---pantolactone,r-pantolactone,d-pantolactone,r-pantoyl lactone,d---pantoyl lactone,pantothenic lactone,r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one,unii-j288d7o0js,2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r,r---pantolactone
Synonym d---pantolactone,r-pantolactone,d-pantolactone,r-pantoyl lactone,d---pantoyl lactone,pantothenic lactone,r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one,unii-j288d7o0js,2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r,r---pantolactone
Synonym d---pantolactone,r-pantolactone,d-pantolactone,r-pantoyl lactone,d---pantoyl lactone,pantothenic lactone,r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one,unii-j288d7o0js,2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r,r---pantolactone
Synonym d---pantolactone,r-pantolactone,d-pantolactone,r-pantoyl lactone,d---pantoyl lactone,pantothenic lactone,r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one,unii-j288d7o0js,2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r,r---pantolactone
Synonym d---pantolactone,r-pantolactone,d-pantolactone,r-pantoyl lactone,d---pantoyl lactone,pantothenic lactone,r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one,unii-j288d7o0js,2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r,r---pantolactone
Synonym d---pantolactone,r-pantolactone,d-pantolactone,r-pantoyl lactone,d---pantoyl lactone,pantothenic lactone,r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one,unii-j288d7o0js,2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r,r---pantolactone
Synonym d---pantolactone,r-pantolactone,d-pantolactone,r-pantoyl lactone,d---pantoyl lactone,pantothenic lactone,r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one,unii-j288d7o0js,2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r,r---pantolactone
Synonym d---pantolactone,r-pantolactone,d-pantolactone,r-pantoyl lactone,d---pantoyl lactone,pantothenic lactone,r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one,unii-j288d7o0js,2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r,r---pantolactone
Synonym d---pantolactone,r-pantolactone,d-pantolactone,r-pantoyl lactone,d---pantoyl lactone,pantothenic lactone,r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one,unii-j288d7o0js,2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r,r---pantolactone
Solubility Information Solubility in water: soluble.
InChI Key SERHXTVXHNVDKA-BYPYZUCNSA-N
SMILES CC1(COC(=O)C1O)C
IUPAC Name (3R)-3-hydroxy-4,4-dimethyloxolan-2-one
Molecular Weight (g/mol) 130.14
PubChem CID 439368
ChEBI CHEBI:16719
Formula Weight 130.14
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Alternative Products
Documentation
Technical documents
Safety and Handling
[D(-)-Pantolactone]
[Warning]
[{code=EYE DAM. 2, sequence=1, description=Serious eye damage/eye irritation Category 2}]
[{code=H319, sequence=1, description=Causes serious eye irritation.}]
[{code=P261, sequence=50, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/.}, {code=P305+P351+P338, sequence=53, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Product Identifier
  • D(-)-Pantolactone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319- Causes serious eye irritation.
Precautionary Statement
  • P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352- IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.