Use promo code "25339" to get your promotional price.
Copper(II) sulfate pentahydrate, 98+%, ACS reagent
Use promo code "25339" to get your promotional price.
| Quantity | 500 g |
|---|---|
| Packaging | Plastic bottle |
??product.attributeSet.Chemical data??
| CAS | 7758-99-8 |
|---|---|
| Molecular Formula | CuH10O9S |
| Molecular Weight (g/mol) | 249.68 |
| MDL Number | MFCD00149681 |
| InChI Key | JZCCFEFSEZPSOG-UHFFFAOYSA-L |
| Synonym | copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas |
| PubChem CID | 24463 |
| ChEBI | CHEBI:31440 |
| SMILES | O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
Specification
| Chemical Name or Material | Copper(II) sulfate pentahydrate |
|---|---|
| Melting Point | 110.0°C |
| CAS | 7758-99-8 |
| Color | Blue |
| Physical Form | Fine Crystals |
| Quantity | 500 g |
| Assay Percent Range | 98.0 to 102% (Iodometry) |
| Molecular Formula | CuH10O9S |
| Linear Formula | CuSO4·5H2O |
| MDL Number | MFCD00149681 |
| Fieser | 01,164; 02,89; 05,162; 06,141; 14,267 |
| Merck Index | 15,2643 |
| Synonym | copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas |
| Synonym | copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas |
| Synonym | copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas |
| Synonym | copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas |
| Synonym | copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas |
| Synonym | copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas |
| Synonym | copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas |
| Synonym | copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas |
| Synonym | copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas |
| Synonym | copper ii sulfate pentahydrate,copper sulfate pentahydrate,cupric sulfate pentahydrate,blue vitriol,calcanthite,bluestone,copper 2+ sulfate pentahydrate,triangle,vencedor,blue copperas |
| Solubility Information | Solubility in water: 320g/L (20°C). Other solubilities: soluble in methanol, slightly soluble in alcohol and glycerol, insoluble in ethanol |
| InChI Key | JZCCFEFSEZPSOG-UHFFFAOYSA-L |
| SMILES | O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
| IUPAC Name | copper;sulfate;pentahydrate |
| Molecular Weight (g/mol) | 249.68 |
| PubChem CID | 24463 |
| ChEBI | CHEBI:31440 |
| Formula Weight | 249.68 |
| Percent Purity | ≥98% |
| Grade | ACS Reagent |
| Packaging | Plastic bottle |
Technical documents
[Copper(II) sulfate pentahydrate]
[Warning]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=SKIN IRRIT. 2, sequence=2, description=Skin corrosion/irritation Category 2}, {code=EYE DAM. 2, sequence=3, description=Serious eye damage/eye irritation Category 2}, {code=AQUATIC ACUTE 1, sequence=4, description=ACUTE AQUATIC Acute 1}, {code=AQUA CHRONIC 1, sequence=5, description=LONG-TERM AQUATIC HAZARD Chronic 1}]
[{code=H302, sequence=1, description=Harmful if swallowed.}, {code=H315, sequence=2, description=Causes skin irritation.}, {code=H319, sequence=3, description=Causes serious eye irritation.}, {code=H410, sequence=4, description=Very toxic to aquatic life with long lasting effects.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P305+P351+P338, sequence=53, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P362, sequence=54, description=Take off contaminated clothing.}, {code=P273, sequence=55, description=Avoid release to the environment.}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}, {code=GHS09, sequence=2, description=ENVIRONMENT}]
Product Identifier
- Copper(II) sulfate pentahydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302- Harmful if swallowed.
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
- H410- Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362- Take off contaminated clothing.
- P273- Avoid release to the environment.