4-Methyl-3-nitrophenol, 98%
CAS: 2042-14-0 | C7H7NO3 | 153.137 g/mol
Additional Details:
Additional Details: CAS Number: 2042-14-0 Weight: 0.05000kg Transport: UN number : 2446 Chem class : 6.1 Pack group : III
Quantity 50 g
Disclaimers: RUO – Research Use Only
Product Code. 11309828
Quantity Price
1 228.0  EUR / 50g
Estimated Shipment
23-11-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
??product.attributeSet.Chemical data??
CAS 2042-14-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007244
InChI Key BQEXDUKMTVYBRK-UHFFFAOYSA-N
Synonym 3-nitro-4-methylphenol,3-nitro-p-cresol,4-methyl-3-nitro-phenol,4-hydroxy-2-nitrotoluene,2-nitro-4-hydroxytoluene,phenol, 4-methyl-3-nitro,p-cresol, 3-nitro,unii-4b422hud6h,4-methyl-5-nitrophenol
PubChem CID 16271
IUPAC Name 4-methyl-3-nitrophenol
SMILES CC1=C(C=C(C=C1)O)[N+](=O)[O-]
Specification
Chemical Name or Material 4-Methyl-3-nitrophenol
CAS 2042-14-0
Melting Point 77°C to 81°C
Odor Phenol-like
Molecular Formula C7H7NO3
MDL Number MFCD00007244
Quantity 50 g
UN Number UN2446
Synonym 3-nitro-4-methylphenol,3-nitro-p-cresol,4-methyl-3-nitro-phenol,4-hydroxy-2-nitrotoluene,2-nitro-4-hydroxytoluene,phenol, 4-methyl-3-nitro,p-cresol, 3-nitro,unii-4b422hud6h,4-methyl-5-nitrophenol
Synonym 3-nitro-4-methylphenol,3-nitro-p-cresol,4-methyl-3-nitro-phenol,4-hydroxy-2-nitrotoluene,2-nitro-4-hydroxytoluene,phenol, 4-methyl-3-nitro,p-cresol, 3-nitro,unii-4b422hud6h,4-methyl-5-nitrophenol
Synonym 3-nitro-4-methylphenol,3-nitro-p-cresol,4-methyl-3-nitro-phenol,4-hydroxy-2-nitrotoluene,2-nitro-4-hydroxytoluene,phenol, 4-methyl-3-nitro,p-cresol, 3-nitro,unii-4b422hud6h,4-methyl-5-nitrophenol
Synonym 3-nitro-4-methylphenol,3-nitro-p-cresol,4-methyl-3-nitro-phenol,4-hydroxy-2-nitrotoluene,2-nitro-4-hydroxytoluene,phenol, 4-methyl-3-nitro,p-cresol, 3-nitro,unii-4b422hud6h,4-methyl-5-nitrophenol
Synonym 3-nitro-4-methylphenol,3-nitro-p-cresol,4-methyl-3-nitro-phenol,4-hydroxy-2-nitrotoluene,2-nitro-4-hydroxytoluene,phenol, 4-methyl-3-nitro,p-cresol, 3-nitro,unii-4b422hud6h,4-methyl-5-nitrophenol
Synonym 3-nitro-4-methylphenol,3-nitro-p-cresol,4-methyl-3-nitro-phenol,4-hydroxy-2-nitrotoluene,2-nitro-4-hydroxytoluene,phenol, 4-methyl-3-nitro,p-cresol, 3-nitro,unii-4b422hud6h,4-methyl-5-nitrophenol
Synonym 3-nitro-4-methylphenol,3-nitro-p-cresol,4-methyl-3-nitro-phenol,4-hydroxy-2-nitrotoluene,2-nitro-4-hydroxytoluene,phenol, 4-methyl-3-nitro,p-cresol, 3-nitro,unii-4b422hud6h,4-methyl-5-nitrophenol
Synonym 3-nitro-4-methylphenol,3-nitro-p-cresol,4-methyl-3-nitro-phenol,4-hydroxy-2-nitrotoluene,2-nitro-4-hydroxytoluene,phenol, 4-methyl-3-nitro,p-cresol, 3-nitro,unii-4b422hud6h,4-methyl-5-nitrophenol
Synonym 3-nitro-4-methylphenol,3-nitro-p-cresol,4-methyl-3-nitro-phenol,4-hydroxy-2-nitrotoluene,2-nitro-4-hydroxytoluene,phenol, 4-methyl-3-nitro,p-cresol, 3-nitro,unii-4b422hud6h,4-methyl-5-nitrophenol
Solubility Information Slightly soluble in water.
InChI Key BQEXDUKMTVYBRK-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)O)[N+](=O)[O-]
IUPAC Name 4-methyl-3-nitrophenol
Molecular Weight (g/mol) 153.137
PubChem CID 16271
Formula Weight 153.14
Percent Purity 98%
Physical Form Powder