4-Methyl-1-pentene, 97%
CAS: 691-37-2 | C6H12 | 84.16 g/mol
Additional Details:
CAS Number:
691-37-2
Weight:
0.01650kg
Transport:
UN number : 3295
Chem class : 3
Pack group : II
| Quantity | 25 mL |
|---|---|
| Packaging | Glass bottle |
Disclaimers:
RUO – Research Use Only
Specification
| Chemical Name or Material | 4-Methyl-1-pentene |
|---|---|
| Name Note | 97% |
| CAS | 691-37-2 |
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | -155°C |
| Color | Colorless |
| Density | 0.6600g/mL |
| Boiling Point | 53°C to 54°C |
| Flash Point | -31°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass bottle |
| Molecular Formula | C6H12 |
| Linear Formula | (CH3)2CHCH2CH=CH2 |
| Refractive Index | 1.3810 to 1.383 |
| MDL Number | MFCD00008949 |
| Quantity | 25 mL |
| Specific Gravity | 0.66 |
| Synonym | 4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer |
| Synonym | 4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer |
| Synonym | 4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer |
| Synonym | 4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer |
| Synonym | 4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer |
| Synonym | 4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer |
| Synonym | 4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer |
| Synonym | 4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer |
| Synonym | 4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer |
| Synonym | 4-methyl-1-pentene,1-pentene, 4-methyl,poly 4-methyl-1-pentene,unii-x10hrj2y7w,x10hrj2y7w,poly 4-methylpentene,isobutylethene,isohexene,4-methylpentene-1,1-pentene, 4-methyl-, homopolymer |
| InChI Key | WSSSPWUEQFSQQG-UHFFFAOYSA-N |
| SMILES | CC(C)CC=C |
| IUPAC Name | 4-methylpent-1-ene |
| Molecular Weight (g/mol) | 84.16 |
| PubChem CID | 12724 |
| Formula Weight | 84.15 |
| Percent Purity | 97% |
| Physical Form | Liquid |
[4-Methyl-1-pentene]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}]
[{code=P210, sequence=50, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}]
[{code=GHS02, sequence=1, description=FLAME}]
Product Identifier
- 4-Methyl-1-pentene
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225- Highly flammable liquid and vapour.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.