1-Tetralone, 97%
CAS: 529-34-0 | C10H10O | 146.19 g/mol
Additional Details:
CAS Number:
529-34-0
Weight:
0.10000kg
| Quantity | 100 g |
|---|
Disclaimers:
RUO – Research Use Only
??product.attributeSet.Chemical data??
| CAS | 529-34-0 |
|---|---|
| Molecular Formula | C10H10O |
| Molecular Weight (g/mol) | 146.19 |
| MDL Number | MFCD00001688 |
| InChI Key | XHLHPRDBBAGVEG-UHFFFAOYSA-N |
| Synonym | alpha-tetralone,1-tetralone,3,4-dihydronaphthalen-1 2h-one,1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-1 2h-naphthalenone,tetralone,a-tetralone,1-oxotetralin,1 2h-naphthalenone, 3,4-dihydro,.alpha.-tetralone |
| PubChem CID | 10724 |
| IUPAC Name | 3,4-dihydro-2H-naphthalen-1-one |
| SMILES | O=C1CCCC2=CC=CC=C12 |
Specification
| Chemical Name or Material | 1-Tetralone |
|---|---|
| CAS | 529-34-0 |
| Melting Point | 6°C to 8°C |
| Density | 1.097 |
| Boiling Point | 257°C |
| Flash Point | 129°C (264°F) |
| Molecular Formula | C10H10O |
| Refractive Index | 1.5695 |
| MDL Number | MFCD00001688 |
| Quantity | 100 g |
| Beilstein | 607374 |
| Synonym | alpha-tetralone,1-tetralone,3,4-dihydronaphthalen-1 2h-one,1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-1 2h-naphthalenone,tetralone,a-tetralone,1-oxotetralin,1 2h-naphthalenone, 3,4-dihydro,.alpha.-tetralone |
| Synonym | alpha-tetralone,1-tetralone,3,4-dihydronaphthalen-1 2h-one,1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-1 2h-naphthalenone,tetralone,a-tetralone,1-oxotetralin,1 2h-naphthalenone, 3,4-dihydro,.alpha.-tetralone |
| Synonym | alpha-tetralone,1-tetralone,3,4-dihydronaphthalen-1 2h-one,1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-1 2h-naphthalenone,tetralone,a-tetralone,1-oxotetralin,1 2h-naphthalenone, 3,4-dihydro,.alpha.-tetralone |
| Synonym | alpha-tetralone,1-tetralone,3,4-dihydronaphthalen-1 2h-one,1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-1 2h-naphthalenone,tetralone,a-tetralone,1-oxotetralin,1 2h-naphthalenone, 3,4-dihydro,.alpha.-tetralone |
| Synonym | alpha-tetralone,1-tetralone,3,4-dihydronaphthalen-1 2h-one,1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-1 2h-naphthalenone,tetralone,a-tetralone,1-oxotetralin,1 2h-naphthalenone, 3,4-dihydro,.alpha.-tetralone |
| Synonym | alpha-tetralone,1-tetralone,3,4-dihydronaphthalen-1 2h-one,1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-1 2h-naphthalenone,tetralone,a-tetralone,1-oxotetralin,1 2h-naphthalenone, 3,4-dihydro,.alpha.-tetralone |
| Synonym | alpha-tetralone,1-tetralone,3,4-dihydronaphthalen-1 2h-one,1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-1 2h-naphthalenone,tetralone,a-tetralone,1-oxotetralin,1 2h-naphthalenone, 3,4-dihydro,.alpha.-tetralone |
| Synonym | alpha-tetralone,1-tetralone,3,4-dihydronaphthalen-1 2h-one,1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-1 2h-naphthalenone,tetralone,a-tetralone,1-oxotetralin,1 2h-naphthalenone, 3,4-dihydro,.alpha.-tetralone |
| Synonym | alpha-tetralone,1-tetralone,3,4-dihydronaphthalen-1 2h-one,1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-1 2h-naphthalenone,tetralone,a-tetralone,1-oxotetralin,1 2h-naphthalenone, 3,4-dihydro,.alpha.-tetralone |
| Synonym | alpha-tetralone,1-tetralone,3,4-dihydronaphthalen-1 2h-one,1,2,3,4-tetrahydronaphthalen-1-one,3,4-dihydro-1 2h-naphthalenone,tetralone,a-tetralone,1-oxotetralin,1 2h-naphthalenone, 3,4-dihydro,.alpha.-tetralone |
| InChI Key | XHLHPRDBBAGVEG-UHFFFAOYSA-N |
| SMILES | O=C1CCCC2=CC=CC=C12 |
| IUPAC Name | 3,4-dihydro-2H-naphthalen-1-one |
| Molecular Weight (g/mol) | 146.19 |
| PubChem CID | 10724 |
| Formula Weight | 146.19 |
| Percent Purity | 97% |