Vacuum Pumps
Reliable vacuum performance is critical for applications such as filtration, rotary evaporation, concentration, aspiration and sample transfer. This vacuum pumps category brings together a broad portfolio of oil-free diaphragm pumps, rotary vane pumps, chemistry vacuum systems and integrated aspiration units designed for demanding laboratory workflows.
Useful Links
Save Now - Exclusive Deals
Product Code 10287097
Product Code 15134816
Product Code 16234611
Product Code 10060322
Product Code 15348191
Product Code 12625635
Product Code 10396852
Product Code 16224611
Product Code 10052433
Product Code 12991131
Product Code 10191942
Product Code 16244611
Product Code 13129065
Product Code 15168886
Product Code 17034237
Must Have
Product Code 10302120
Product Code 10255919
Product Code 17225123
Product Code 16211762
Product Code 16676895
Product Code 15484590
Product Code 15567479
Product Code 17724919
Product Code 11986858
Product Code 13492059
Product Code 10238232
Product Code 10641120
Product Code 10450031
Product Code 16264611
FAQ
For routine filtration, oil-free diaphragm vacuum pumps from brands such as Fisherbrand, KNF and Vacuubrand are typically recommended. They provide sufficient vacuum for membrane filtration, low maintenance operation and reduced contamination risk compared with oil-sealed rotary vane pumps, making them ideal for everyday laboratory workflows and student environments.
Diaphragm pumps from Fisherbrand, KNF or Cole Parmer are ideal for chemically aggressive or aqueous samples, offering oil-free operation and easy maintenance. Rotary vane pumps from Fisherbrand, Welch Ilmvac or Büchi deliver deeper vacuum for freeze-drying, Schlenk lines or high-boiling solvents, but require oil changes and more attention to exhaust management.
For rotary evaporators and solvent evaporation, look for chemistry-resistant diaphragm or rotary vane pumps from Vacuubrand, Büchi or Welch Ilmvac. Key specifications include ultimate vacuum, flow rate, corrosion-resistant wetted parts, optional speed control and compatibility with vacuum controllers to prevent bumping, improve solvent recovery and protect the pump from condensate.
Chemical-resistant vacuum pumps from Fisherbrand, Vacuubrand, KNF and Cole Parmer are designed to handle corrosive vapours from acids, bases and organic solvents. Enhanced materials in diaphragms, valves and housings help maintain performance, extend pump lifetime and reduce unplanned downtime, especially in analytical, synthesis and sample preparation laboratories.
To minimise maintenance, select oil-free diaphragm pumps from brands such as Fisherbrand, KNF or Vacuubrand where possible, use appropriate filtration or cold traps, and operate within recommended pressure and temperature limits. Regular checks of tubing, connections and exhaust filters, plus scheduled servicing, help maintain stable vacuum and extend pump service life.