accessibility menu, dialog, popup

UserName

CAS RN 999-81-5

ClCH3NCH3CH3Cl

CAS RN 999-81-5

IUPAC Name: (2-chloroethyl)trimethylazanium chloride
Synonyms: chlormequat chloride
Molecular Weight (g/mol): 158.07
Molecular Formula: C5H13Cl2N
InChi Key: UHZZMRAGKVHANO-UHFFFAOYSA-M
SMILES: [Cl-].C[N+](C)(C)CCCl
Molecular Weight (g/mol) 
  • (7)
  • (5)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
Concentration 
  • (2)
  • (2)
Solvent or Matrix 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (7)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)