accessibility menu, dialog, popup

UserName

CAS RN 999-55-3

H2COOCH2

CAS RN 999-55-3

IUPAC Name: prop-2-en-1-yl prop-2-enoate
Synonyms: prop-2-en-1-yl prop-2-enoate
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: QTECDUFMBMSHKR-UHFFFAOYSA-N
SMILES: C=CCOC(=O)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results