accessibility menu, dialog, popup

UserName

CAS RN 99817-36-4

H3CClOClNOO

CAS RN 99817-36-4

IUPAC Name: 2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate
Synonyms: 2,4-dichloro-3-ethyl-6-nitrobenzenolate
Molecular Weight (g/mol): 235.04
Molecular Formula: C8H6Cl2NO3
InChi Key: YTVCECQSAPGJBB-UHFFFAOYSA-M
SMILES: CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)