accessibility menu, dialog, popup

UserName

CAS RN 99514-85-9

NHONOOBr

CAS RN 99514-85-9

IUPAC Name: N-(2-bromophenyl)-4-nitrobenzamide
Synonyms: N-(2-bromophenyl)-4-nitrobenzamide
Molecular Weight (g/mol): 321.13
Molecular Formula: C13H9BrN2O3
InChi Key: FRFOXBSTPQRRJL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C(=O)NC1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results