accessibility menu, dialog, popup

UserName

CAS RN 99-98-9

IUPAC Name: N1,N1-dimethylbenzene-1,4-diamine
Synonyms: dimethyl-p-phenylenediamine DMPD N,N-dimethyl phenylene diamine dimethyl-para-phenylenediamine N,N-dimethyl-p-fenylendiamin dimethyl-4-phenylenediamine dimethyl 1,4-phenylenediamine n,n-dimethyl phenylene diamine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: BZORFPDSXLZWJF-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (7)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (3)
  • (4)
  • (4)