accessibility menu, dialog, popup

UserName

H3CCH3CH3

CAS RN 99-85-4

IUPAC Name: 1-methyl-4-(propan-2-yl)cyclohexa-1,4-diene
Synonyms: α terpinene moslene gamma-terpinene terpinene, α gamma terpinene gamma-terpinen crithmene gamma-terpinene (natural) 1,4-p-menthadiene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: YKFLAYDHMOASIY-UHFFFAOYSA-N
SMILES: CC(C)C1=CCC(C)=CC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results