accessibility menu, dialog, popup

UserName

CAS RN 99-08-1

CH3NOO

CAS RN 99-08-1

IUPAC Name: 1-methyl-3-nitrobenzene
Synonyms: 3-nitrotoluene
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: QZYHIOPPLUPUJF-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results