accessibility menu, dialog, popup

UserName

H3CNCH3HO

CAS RN 99-07-0

IUPAC Name: 3-(dimethylamino)phenol
Synonyms: phenol, 3-(dimethylamino)-
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: MESJRHHDBDCQTH-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC(O)=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)