accessibility menu, dialog, popup

UserName

CH3NH3CCH3OPOOOPHOOO(R)O(R)(R)HO(S)HONNONH2

CAS RN 987-78-0

IUPAC Name: {2-[({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]ethyl}trimethylazanium
Synonyms: citicoline cdp-colina citicolinum citicolina citidin difosfato de colina 胞磷胆碱 cytidindiphosphocholin cdp-choline
Molecular Weight (g/mol): 488.33
Molecular Formula: C14H26N4O11P2
InChi Key: RZZPDXZPRHQOCG-OJAKKHQRSA-N
SMILES: C[N+](C)(C)CCOP([O-])(=O)OP(O)(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)